accessibility menu, dialog, popup

UserName

CAS RN 41037-26-7

CH3NOCH3

CAS RN 41037-26-7

IUPAC Name: 6-methoxy-4-methylquinoline
Synonyms: 6-methoxy-4-methylquinoline
Molecular Weight (g/mol): 173.22
Molecular Formula: C11H11NO
InChi Key: MBVGYFIYXWVPQY-UHFFFAOYSA-N
SMILES: COC1=CC=C2N=CC=C(C)C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results