missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-4-methylquinoline Hydrate 98.0+%, TCI America™

Catalog No. M10875G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10875G 5 g
M10871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M10875G Supplier TCI America Supplier No. M10875G
Only null left

Chemical Identifiers

CAS 41037-26-7
Molecular Formula C11H11NO
Molecular Weight (g/mol) 173.215
MDL Number MFCD00191547
InChI Key MBVGYFIYXWVPQY-UHFFFAOYSA-N
PubChem CID 258761
IUPAC Name 6-methoxy-4-methylquinoline
SMILES CC1=C2C=C(C=CC2=NC=C1)OC

Specifications

Melting Point 53°C
Color White-Yellow
Boiling Point 158°C
Quantity 5 g
Formula Weight 173.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methoxy-4-methylquinoline Hydrate