missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Methoxy-4-methylquinoline Hydrate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 53°C |
| Color | White-Yellow |
| Boiling Point | 158°C |
| Quantity | 1 g |
| Formula Weight | 173.22 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 6-Methoxy-4-methylquinoline Hydrate |
Safety and Handling
| TSCA | No |