missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-2-naphthylglyoxal hydrate, 98%, dry wt. basis

Catalog No. AAL2026006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2026006 5 g
AAL2026003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2026006 Supplier Thermo Scientific Chemicals Supplier No. L2026006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 745783-88-4 | C13H12O4 | 232.235 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 745783-88-4
Molecular Formula C13H12O4
Molecular Weight (g/mol) 232.235
MDL Number MFCD05664091
InChI Key CBFAHRRUWMTAQH-UHFFFAOYSA-N
Synonym 6-methoxynaphthylglyoxal hydrate, 2-6-methoxynaphthalen-1-yl-2-oxoacetaldehyde hydrate, 2-6-methoxy-1-naphthyl-2-oxo-acetaldehyde, pubchem12296, 2-6-methoxynaphthyl-2-oxoethanal, hydrate, 2-6-methoxy-1-naphthalenyl-2-oxoacetaldehyde hydrate, 6-methoxynaphthalen-1-yl oxo acetaldehyde-water 1/1, 2-6-methoxynaphthalen-1-yl-2-oxidanylidene-ethanal hydrate
PubChem CID 44118759
IUPAC Name 2-(6-methoxynaphthalen-1-yl)-2-oxoacetaldehyde;hydrate
SMILES COC1=CC2=C(C=C1)C(=CC=C2)C(=O)C=O.O

Specifications

Melting Point 125°C to 130°C
Quantity 5 g
Formula Weight 214.22 (Anhydrous)
Percent Purity 98%
Assay (dry wt.)
Chemical Name or Material 6-Methoxy-2-naphthylglyoxal hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only