accessibility menu, dialog, popup

UserName

CAS RN 745783-88-4

IUPAC Name: 2-(6-methoxynaphthalen-2-yl)-2-oxoacetaldehyde
Synonyms: 2-(6-methoxynaphthalen-2-yl)-2-oxoacetaldehyde
Molecular Weight (g/mol): 214.22
Molecular Formula: C13H10O3
InChi Key: BAVQWNGBJSPFBJ-UHFFFAOYSA-N
SMILES: COC1=CC2=CC=C(C=C2C=C1)C(=O)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results