missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-2-naphthoic acid, 98+%

Catalog No. AAL1461003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1461003 1 g
AAL1461006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1461003 Supplier Thermo Scientific Chemicals Supplier No. L1461003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2471-70-7 | C12H9O3 | 201.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2471-70-7
Molecular Formula C12H9O3
Molecular Weight (g/mol) 201.20
MDL Number MFCD00092750
InChI Key YZBILXXOZFORFE-UHFFFAOYSA-M
Synonym 6-methoxy-2-naphthoic acid, 2-naphthalenecarboxylic acid, 6-methoxy, unii-65ih1sv66a, 6-methoxy-2-naphthoicacid, 6-methoxy-2-naphthalenecarboxylic acid, 6-methoxy-naphthalene-2-carboxylic acid, naproxen specified impurity o ep, naproxen impurity o, 2-methoxy-6-naphthalenecarboxylic acid, zlchem 1096
PubChem CID 349181
IUPAC Name 6-methoxynaphthalene-2-carboxylate
SMILES COC1=CC=C2C=C(C=CC2=C1)C([O-])=O

Specifications

Melting Point 192°C to 194°C
Quantity 1 g
Beilstein 2211233
Formula Weight 202.21
Percent Purity ≥98%
Chemical Name or Material 6-Methoxy-2-naphthoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only