missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-1-tetralone, 99%

Catalog No. p-3778995
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC126120250 25 g Glass bottle
AC126121000 100 g Glass bottle
AC126125000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC126120250 Supplier Thermo Scientific Chemicals Supplier No. 126120250
Only null left

CAS: 1078-19-9 | C11H12O2 | 176.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1078-19-9
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.21
MDL Number MFCD00001695
InChI Key MNALUTYMBUBKNX-UHFFFAOYSA-N
Synonym 6-methoxy-1-tetralone, 6-methoxy-3,4-dihydronaphthalen-1 2h-one, 6-methoxytetralone, 6-methoxy-alpha-tetralone, 1 2h-naphthalenone, 3,4-dihydro-6-methoxy, 6-methoxy-1,2,3,4-tetrahydronaphthalen-1-one, 6-methoxy-3,4-dihydro-1 2h-naphthalenone, 6-methoxytetralin-1-one, 3,4-dihydro-6-methoxy-1 2h-naphthalenone, 6-methoxy-.alpha.-tetralone
PubChem CID 14112
IUPAC Name 6-methoxy-3,4-dihydro-2H-naphthalen-1-one
SMILES COC1=CC2=C(C=C1)C(=O)CCC2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 77.0°C to 79.0°C
Color Brown to Yellow
Boiling Point 171.0°C (11.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Beilstein 09, II, 889
Merck Index 15, 6074
Formula Weight 176.21
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 6-Methoxy-1-tetralone
Show More Show Less
EINECSNumber 214-078-

RUO – Research Use Only