missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-1-tetralone, 99%

Catalog No. AAA1596706
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1596706 5 g
AAA1596714 25 g
AAA1596722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1596706 Supplier Thermo Scientific Chemicals Supplier No. A1596706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1078-19-9 | C11H12O2 | 176.215 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1078-19-9
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD00001695
InChI Key MNALUTYMBUBKNX-UHFFFAOYSA-N
Synonym 6-methoxy-1-tetralone, 6-methoxy-3,4-dihydronaphthalen-1 2h-one, 6-methoxytetralone, 6-methoxy-alpha-tetralone, 1 2h-naphthalenone, 3,4-dihydro-6-methoxy, 6-methoxy-1,2,3,4-tetrahydronaphthalen-1-one, 6-methoxy-3,4-dihydro-1 2h-naphthalenone, 6-methoxytetralin-1-one, 3,4-dihydro-6-methoxy-1 2h-naphthalenone, 6-methoxy-.alpha.-tetralone
PubChem CID 14112
IUPAC Name 6-methoxy-3,4-dihydro-2H-naphthalen-1-one
SMILES COC1=CC2=C(C=C1)C(=O)CCC2

Specifications

Melting Point 77°C to 80°C
Boiling Point 171°C (11 mmHg)
Quantity 5 g
Beilstein 972739
Merck Index 14,6000
Formula Weight 176.22
Percent Purity 99%
Chemical Name or Material 6-Methoxy-1-tetralone
EINECSNumber 214-078-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only