missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-1-indanone 98.0+%, TCI America™

Catalog No. M15355G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M15355G 5 g
M153525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M15355G Supplier TCI America Supplier No. M15355G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13623-25-1
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00021232
InChI Key UJGDLLGKMWVCPT-UHFFFAOYSA-N
Synonym 6-methoxy-1-indanone, 6-methoxy-1h-indanone, 6-methoxy-indan-1-one, 6-methoxy-2,3-dihydro-1h-inden-1-one, 6-methoxyindan-1-one, 2,3-dihydro-6-methoxyinden-1-one, 1h-inden-1-one, 2,3-dihydro-6-methoxy, 6-methoxyindanone, 6-methoxy indanone, 6-methoxy-1-indanon
PubChem CID 334036
IUPAC Name 6-methoxy-2,3-dihydro-1H-inden-1-one
SMILES COC1=CC=C2CCC(=O)C2=C1

Specifications

Melting Point 109°C
Color Yellow
Quantity 5 g
Formula Weight 162.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methoxy-1-indanone