accessibility menu, dialog, popup

UserName

CH3OO

CAS RN 13623-25-1

IUPAC Name: 6-methoxy-2,3-dihydro-1H-inden-1-one
Synonyms: 6-methoxy-2,3-dihydroinden-1-one
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: UJGDLLGKMWVCPT-UHFFFAOYSA-N
SMILES: COC1=CC=C2CCC(=O)C2=C1
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)