missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-1-indanone, 99%, Thermo Scientific Chemicals

Catalog No. aal1581906
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1581906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1581906 Supplier Thermo Scientific Chemicals Supplier No. L1581906

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13623-25-1
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00021232
InChI Key UJGDLLGKMWVCPT-UHFFFAOYSA-N
Synonym 6-methoxy-1-indanone, 6-methoxy-1h-indanone, 6-methoxy-indan-1-one, 6-methoxy-2,3-dihydro-1h-inden-1-one, 6-methoxyindan-1-one, 2,3-dihydro-6-methoxyinden-1-one, 1h-inden-1-one, 2,3-dihydro-6-methoxy, 6-methoxyindanone, 6-methoxy indanone, 6-methoxy-1-indanon
PubChem CID 334036
IUPAC Name 6-methoxy-2,3-dihydroinden-1-one
SMILES COC1=CC=C2CCC(=O)C2=C1

Specifications

Melting Point 104°C to 109°C
Quantity 5 g
Beilstein 1238602
Formula Weight 162.19
Percent Purity 99%
Chemical Name or Material 6-Methoxy-1-indanone
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only