missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-1,2,3,4-tetrahydroquinoline, 97%

CAS: 120-15-0 | C10H13NO | 163.22 g/mol

Supplier:  Thermo Scientific Chemicals H6300206

 View more versions of this product

Catalog No. AAH6300206


Only null left
Explore available promotions

Description

Description

1,2,3,4-Tetrahydro-6-methoxyquinoline is used as chemical reagents, organic intermediates, fine chemicals, pharmaceutical research and development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3,4-Tetrahydro-6-methoxyquinoline is used as chemical reagents, organic intermediates, fine chemicals, pharmaceutical research and development.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

6-Methoxy-1,2,3,4-tetrahydroquinoline
34°C to 37°C
MFCD00090494
thallin, thalline, 6-methoxy-1,2,3,4-tetrahydro-quinoline, 6-methoxytetrahydroquinoline, quinoline, 1,2,3,4-tetrahydro-6-methoxy, 1,2,3,4-tetrahydro-6-methoxyquinoline, methyl 1,2,3,4-tetrahydro-6-quinolyl ether, quinoline l,2,3,4-tetrahydro-6-methoxy, wln: t66 bmt & j ho1, 1,3,4-tetrahydro-6-methoxyquinoline
COC1=CC2=C(C=C1)NCCC2
163.22
163.22
120-15-0
C10H13NO
5 g
FRXSZNDVFUDTIR-UHFFFAOYSA-N
6-methoxy-1,2,3,4-tetrahydroquinoline
67113
97%

Chemical Identifiers

120-15-0
163.22
FRXSZNDVFUDTIR-UHFFFAOYSA-N
67113
COC1=CC2=C(C=C1)NCCC2
C10H13NO
MFCD00090494
thallin, thalline, 6-methoxy-1,2,3,4-tetrahydro-quinoline, 6-methoxytetrahydroquinoline, quinoline, 1,2,3,4-tetrahydro-6-methoxy, 1,2,3,4-tetrahydro-6-methoxyquinoline, methyl 1,2,3,4-tetrahydro-6-quinolyl ether, quinoline l,2,3,4-tetrahydro-6-methoxy, wln: t66 bmt & j ho1, 1,3,4-tetrahydro-6-methoxyquinoline
6-methoxy-1,2,3,4-tetrahydroquinoline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 204-374-8

RTECSNumber : VC2940000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only