missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-1,2,3,4-tetrahydroquinoline, 97%

Catalog No. p-7041656
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63002MD 250 mg
AAH6300203 1 g
AAH6300206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63002MD Supplier Thermo Scientific Chemicals Supplier No. H63002MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 120-15-0 | C10H13NO | 163.22 g/mol

1,2,3,4-Tetrahydro-6-methoxyquinoline is used as chemical reagents, organic intermediates, fine chemicals, pharmaceutical research and development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3,4-Tetrahydro-6-methoxyquinoline is used as chemical reagents, organic intermediates, fine chemicals, pharmaceutical research and development.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 120-15-0
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
MDL Number MFCD00090494
InChI Key FRXSZNDVFUDTIR-UHFFFAOYSA-N
Synonym thallin, thalline, 6-methoxy-1,2,3,4-tetrahydro-quinoline, 6-methoxytetrahydroquinoline, quinoline, 1,2,3,4-tetrahydro-6-methoxy, 1,2,3,4-tetrahydro-6-methoxyquinoline, methyl 1,2,3,4-tetrahydro-6-quinolyl ether, quinoline l,2,3,4-tetrahydro-6-methoxy, wln: t66 bmt & j ho1, 1,3,4-tetrahydro-6-methoxyquinoline
PubChem CID 67113
IUPAC Name 6-methoxy-1,2,3,4-tetrahydroquinoline
SMILES COC1=CC2=C(C=C1)NCCC2

Specifications

Melting Point 34°C to 37°C
Quantity 250 mg
Formula Weight 163.22
Percent Purity 97%
Chemical Name or Material 6-Methoxy-1,2,3,4-tetrahydroquinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 204-374-8
RTECSNumber VC2940000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only