missing translation for 'onlineSavingsMsg'
Learn More

6-Mercaptopurine Monohydrate 98.0+%, TCI America™

Catalog No. p-7131961
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
M00631G 1 g
M00635G 5 g
2 options

Catalog No. M00631G

Supplier: TCI America M00631G

Only null left

Chemical Identifiers

CAS 6112-76-1
Molecular Formula C5H6N4OS
Molecular Weight (g/mol) 170.19
MDL Number MFCD03854445,MFCD01461928
InChI Key WFFQYWAAEWLHJC-UHFFFAOYSA-N
Synonym 6-mercaptopurine monohydrate, 6-mercaptopurine hydrate, mercaptopurine monohydrate, mercaptopurine hydrate, 6h-purine-6-thione, 1,7-dihydro-, monohydrate, 6,7-dihydro-3h-purine-6-thione hydrate, purine-6-thiol monohydrate, 1,7-dihydro-6h-purine-6-thione monohydrate, purine-6-thiol, monohydrate, unii-e7wed276i5
PubChem CID 2724350
ChEBI CHEBI:31822
IUPAC Name 6,7-dihydro-3H-purine-6-thione hydrate
SMILES O.S=C1N=CNC2=C1NC=N2

Specifications

Color Yellow
Quantity 1 g
Formula Weight 152.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Mercaptopurine Monohydrate
EINECSNumber (9)-1432
RTECSNumber UP0400000
TSCA No