missing translation for 'onlineSavingsMsg'
Learn More

6-Mercaptopurine monohydrate, 98%

Catalog No. p-7081979
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1219703 1 g
AAA1219706 5 g
AAA1219714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1219703 Supplier Thermo Scientific Chemicals Supplier No. A1219703
Only null left

CAS: 6112-76-1 | C5H6N4OS | 170.19 g/mol

6-Mercaptopurine is a widely used antileukemic agent that inhibits de novopurine synthesis through incorporation of thiopurine methyltransferase metabolites into DNA and RNA. It is also an antitumor agent and induces apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Mercaptopurine is a widely used antileukemic agent that inhibits de novopurine synthesis through incorporation of thiopurine methyltransferase metabolites into DNA and RNA. It is also an antitumor agent and induces apoptosis.

Solubility
Soluble in dilute solutions of alkali hydroxides, hot alcohol, and 1 N NaOH (50 mg/ml). Insoluble in water.

Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 6112-76-1
Molecular Formula C5H6N4OS
Molecular Weight (g/mol) 170.19
MDL Number MFCD03854445,MFCD01461928
InChI Key WFFQYWAAEWLHJC-UHFFFAOYSA-N
Synonym 6-mercaptopurine monohydrate, 6-mercaptopurine hydrate, mercaptopurine monohydrate, mercaptopurine hydrate, 6h-purine-6-thione, 1,7-dihydro-, monohydrate, 6,7-dihydro-3h-purine-6-thione hydrate, purine-6-thiol monohydrate, 1,7-dihydro-6h-purine-6-thione monohydrate, purine-6-thiol, monohydrate, unii-e7wed276i5
PubChem CID 2724350
ChEBI CHEBI:31822
IUPAC Name 3,7-dihydropurine-6-thione;hydrate
SMILES O.S=C1N=CNC2=C1NC=N2

Specifications

Melting Point ∼315°C (decomposition)
Quantity 1 g
Beilstein 4012091
Merck Index 14,5871
Solubility Information Soluble in dilute solutions of alkali hydroxides,hot alcohol,and 1 N NaOH (50mg/ml). Insoluble in water.
Formula Weight 170.19 (152.17 Anhydrous)
Percent Purity 98%
Chemical Name or Material 6-Mercaptopurine monohydrate
Hazard Category H302-H315-H319-H335-H341-H361
Hazard Statement GHS H Statement
H341-H361-H302
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Precautionary Statement P202-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
EINECSNumber 200-037-4
RTECSNumber UP0400000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only