missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Maleimidocaproic acid N-succinimidyl ester, 95%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 69°C to 74°C |
| Color | White to Brown |
| Quantity | 250 mg |
| Beilstein | 1499815 |
| Sensitivity | Light and moisture sensitive; Hygroscopic |
| Formula Weight | 308.29 |
| Percent Purity | 95% |
| Physical Form | Solid |
| Chemical Name or Material | 6-Maleimidocaproic acid N-succinimidyl ester |
Safety and Handling
| missing translation for 'healthHazard1' | H315-H319-H335 |
| missing translation for 'healthHazard3' | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| storageNote1 | -30° to -10°C |
| Recommended Storage | Store at -20°C; Store under Nitrogen |
RUO – Research Use Only