accessibility menu, dialog, popup

UserName

CAS RN 55750-63-5

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 6-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanoate
Synonyms: 2,5-dioxopyrrolidin-1-yl 6-(2,5-dioxopyrrol-1-yl)hexanoate
Molecular Weight (g/mol): 308.29
Molecular Formula: C14H16N2O6
InChi Key: VLARLSIGSPVYHX-UHFFFAOYSA-N
SMILES: O=C(CCCCCN1C(=O)C=CC1=O)ON1C(=O)CCC1=O
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)