missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ 6-JOE, SE (6-Carboxy-4',5'-Dichloro-2',7'-Dimethoxyfluorescein, Succinimidyl Ester)

Catalog No. c6171mp
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C6171MP 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C6171MP Supplier Molecular Probes™ Supplier No. C6171MP

6-JOE, SE (6-Carboxy-4',5'-Dichloro-2',7'-Dimethoxyfluorescein, Succinimidyl Ester)

This single isomer preparation of the amine-reactive dye 6-JOE, SE has fluorescence characteristics that are red-shifted as compared to fluorescein (excitation/emission is 522/550 nm) and is one of the traditional fluorophores used in automated DNA sequencing.

Specifications

Quantity 5 mg
Product Type 6-JOE SE
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye JOE
Label Type Classic Dyes
Emission 550
Excitation 522

For Research Use Only. Not for use in diagnostic procedures.