missing translation for 'onlineSavingsMsg'
Learn More

6-Isopropylquinoline 98.0+%, TCI America™

Catalog No. I05295G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I05295G Supplier TCI America Supplier No. I05295G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 135-79-5
Molecular Formula C12H13N
Molecular Weight (g/mol) 171.243
MDL Number MFCD00047615
InChI Key NKCQEIXYLHACJC-UHFFFAOYSA-N
Synonym 6-isopropylquinoline, quinoline, 6-1-methylethyl, 6-propan-2-yl quinoline, unii-8ge3n8wv7y, 6-1-methylethyl quinoline, 8ge3n8wv7y, 6-isopropyl quinoline, pubchem5907, acmc-1bt55, dsstox_cid_27465
PubChem CID 67285
IUPAC Name 6-propan-2-ylquinoline
SMILES CC(C)C1=CC2=C(C=C1)N=CC=C2

Specifications

Color Yellow
Boiling Point 143°C
Quantity 5 g
Formula Weight 171.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 6-Isopropylquinoline