CAS RN 135-79-5
IUPAC Name:
6-(propan-2-yl)quinoline
Synonyms:
6-isopropylquinoline
Molecular Weight (g/mol):
171.24
Molecular Formula:
C12H13N
InChi Key:
NKCQEIXYLHACJC-UHFFFAOYSA-N
SMILES:
CC(C)C1=CC=C2N=CC=CC2=C1
1
–
1
of
1
results
1
6-Isopropylquinoline 98.0+%, TCI America™
CAS: 135-79-5 Molecular Formula: C12H13N Molecular Weight (g/mol): 171.243 MDL Number: MFCD00047615 InChI Key: NKCQEIXYLHACJC-UHFFFAOYSA-N Synonym: 6-isopropylquinoline,quinoline, 6-1-methylethyl,6-propan-2-yl quinoline,unii-8ge3n8wv7y,6-1-methylethyl quinoline,8ge3n8wv7y,6-isopropyl quinoline,pubchem5907,acmc-1bt55,dsstox_cid_27465 PubChem CID: 67285 IUPAC Name: 6-propan-2-ylquinoline SMILES: CC(C)C1=CC2=C(C=C1)N=CC=C2