missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxypyridazine-3-carboxylic Acid Monohydrate 98.0+%, TCI America™

Catalog No. H13871G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13871G 1 g
H13875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13871G Supplier TCI America Supplier No. H13871G

Chemical Identifiers

CAS 306934-80-5
Molecular Formula C5H6N2O4
Molecular Weight (g/mol) 158.113
MDL Number MFCD09064936
InChI Key DZCBVXKRERSHAA-UHFFFAOYSA-N
PubChem CID 2774651
IUPAC Name 6-oxo-1H-pyridazine-3-carboxylic acid;hydrate
SMILES C1=CC(=O)NN=C1C(=O)O.O

Specifications

Melting Point 257°C
Color White
Quantity 1 g
Formula Weight 140.10
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Hydroxypyridazine-3-carboxylic Acid Monohydrate
EINECSNumber (5)-0974
TSCA No