Learn More
6-Hydroxynicotinic acid, 98%
CAS: 5006-66-6 | C6H5NO3 | 139.11 g/mol
Supplier: Thermo Scientific Chemicals A1580214
Description
Metabolite of nicotinamide. Stimulates adrenocortical secretion
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 6-Hydroxynicotinic acid | |
| 314°C to 316°C | |
| MFCD00006277 | |
| 472182 | |
| BLHCMGRVFXRYRN-UHFFFAOYSA-N | |
| 6-oxo-1H-pyridine-3-carboxylic acid | |
| 72924 | |
| 139.11 |
| 5006-66-6 | |
| C6H5NO3 | |
| 25 g | |
| 6-hydroxynicotinic acid, 2-hydroxy-5-pyridinecarboxylic acid, 6-hydroxypyridine-3-carboxylic acid, 6-hydroxy nicotinic acid, 3-pyridinecarboxylic acid, 1,6-dihydro-6-oxo, 2-hydroxypyridine-5-carboxylic acid, 2-pyridone-5-carboxylic acid, 6-hydroxy-nicotinic acid, 6-hydroxynicotinate, 1,6-dihydro-6-oxo-3-pyridinecarboxylic acid | |
| C1=CC(=O)NC=C1C(=O)O | |
| 139.11 | |
| CHEBI:16168 | |
| 98% |
Chemical Identifiers
| 5006-66-6 | |
| 139.11 | |
| BLHCMGRVFXRYRN-UHFFFAOYSA-N | |
| 72924 | |
| 6-oxo-1H-pyridine-3-carboxylic acid |
| C6H5NO3 | |
| MFCD00006277 | |
| 6-hydroxynicotinic acid, 2-hydroxy-5-pyridinecarboxylic acid, 6-hydroxypyridine-3-carboxylic acid, 6-hydroxy nicotinic acid, 3-pyridinecarboxylic acid, 1,6-dihydro-6-oxo, 2-hydroxypyridine-5-carboxylic acid, 2-pyridone-5-carboxylic acid, 6-hydroxy-nicotinic acid, 6-hydroxynicotinate, 1,6-dihydro-6-oxo-3-pyridinecarboxylic acid | |
| CHEBI:16168 | |
| C1=CC(=O)NC=C1C(=O)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 225-682-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only