accessibility menu, dialog, popup

UserName

CAS RN 5006-66-6

IUPAC Name: 6-oxo-1,6-dihydropyridine-3-carboxylic acid
Synonyms: 6-oxo-1H-pyridine-3-carboxylic acid
Molecular Weight (g/mol): 139.11
Molecular Formula: C6H5NO3
InChi Key: BLHCMGRVFXRYRN-UHFFFAOYSA-N
SMILES: OC(=O)C1=CNC(=O)C=C1
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)