missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxynicotinic acid, 98%

Catalog No. AAA1580222
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1580222 100 g
AAA1580206 5 g
AAA1580214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1580222 Supplier Thermo Scientific Chemicals Supplier No. A1580222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5006-66-6 | C6H5NO3 | 139.11 g/mol

Metabolite of nicotinamide. Stimulates adrenocortical secretion

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5006-66-6
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00006277
InChI Key BLHCMGRVFXRYRN-UHFFFAOYSA-N
Synonym 6-hydroxynicotinic acid, 2-hydroxy-5-pyridinecarboxylic acid, 6-hydroxypyridine-3-carboxylic acid, 6-hydroxy nicotinic acid, 3-pyridinecarboxylic acid, 1,6-dihydro-6-oxo, 2-hydroxypyridine-5-carboxylic acid, 2-pyridone-5-carboxylic acid, 6-hydroxy-nicotinic acid, 6-hydroxynicotinate, 1,6-dihydro-6-oxo-3-pyridinecarboxylic acid
PubChem CID 72924
ChEBI CHEBI:16168
IUPAC Name 6-oxo-1H-pyridine-3-carboxylic acid
SMILES C1=CC(=O)NC=C1C(=O)O

Specifications

Melting Point 314°C to 316°C
Quantity 100 g
Beilstein 472182
Formula Weight 139.11
Percent Purity 98%
Chemical Name or Material 6-Hydroxynicotinic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 225-682-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only