missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy-2-naphthonitrile 98.0+%, TCI America™

Catalog No. H09955G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H09955G Supplier TCI America Supplier No. H09955G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 52927-22-7
Molecular Formula C11H7NO
Molecular Weight (g/mol) 169.18
MDL Number MFCD01863480
InChI Key WKTNIBWKHNIPQR-UHFFFAOYSA-N
Synonym 6-Cyano-2-naphthol
PubChem CID 4589476
IUPAC Name 6-hydroxynaphthalene-2-carbonitrile
SMILES OC1=CC2=CC=C(C=C2C=C1)C#N

Specifications

Melting Point 168°C
Quantity 5 g
Formula Weight 169.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Hydroxy-2-naphthonitrile