missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Hydroxy-2-naphthoic Acid 98.0+%, TCI America™
$35.41 - $363.49
Chemical Identifiers
| CAS | 16712-64-4 |
|---|---|
| Molecular Formula | C11H8O3 |
| Molecular Weight (g/mol) | 188.182 |
| MDL Number | MFCD00060070 |
| InChI Key | KAUQJMHLAFIZDU-UHFFFAOYSA-N |
| Synonym | 6-hydroxy-2-naphthoic acid, 2-naphthalenecarboxylic acid, 6-hydroxy, 6-hydroxy-2-naphthalenecarboxylic acid, 6-carboxy-2-naphthol, 2-hydroxynaphthalene-6-carboxylic acid, 6-hydroxy-beta-naphthoic acid, unii-72xa858i6p, 6-hydroxy-naphthalene-2-carboxylic acid, 2-naphthoic acid, 6-hydroxy, 2-hydroxy-6-naphthoic acid |
| PubChem CID | 85557 |
| IUPAC Name | 6-hydroxynaphthalene-2-carboxylic acid |
| SMILES | C1=CC2=C(C=CC(=C2)O)C=C1C(=O)O |
Chemical Identifiers
| 16712-64-4 | |
| 188.182 | |
| KAUQJMHLAFIZDU-UHFFFAOYSA-N | |
| 85557 | |
| C1=CC2=C(C=CC(=C2)O)C=C1C(=O)O |
| C11H8O3 | |
| MFCD00060070 | |
| 6-hydroxy-2-naphthoic acid, 2-naphthalenecarboxylic acid, 6-hydroxy, 6-hydroxy-2-naphthalenecarboxylic acid, 6-carboxy-2-naphthol, 2-hydroxynaphthalene-6-carboxylic acid, 6-hydroxy-beta-naphthoic acid, unii-72xa858i6p, 6-hydroxy-naphthalene-2-carboxylic acid, 2-naphthoic acid, 6-hydroxy, 2-hydroxy-6-naphthoic acid | |
| 6-hydroxynaphthalene-2-carboxylic acid |
Specifications
| 16712-64-4 | |
| C11H8O3 | |
| 5 g | |
| KAUQJMHLAFIZDU-UHFFFAOYSA-N | |
| 6-hydroxynaphthalene-2-carboxylic acid | |
| 85557 | |
| ≥98.0% (GC,T) | |
| 6-Hydroxy-2-naphthoic Acid |
| 248°C | |
| MFCD00060070 | |
| 6-hydroxy-2-naphthoic acid, 2-naphthalenecarboxylic acid, 6-hydroxy, 6-hydroxy-2-naphthalenecarboxylic acid, 6-carboxy-2-naphthol, 2-hydroxynaphthalene-6-carboxylic acid, 6-hydroxy-beta-naphthoic acid, unii-72xa858i6p, 6-hydroxy-naphthalene-2-carboxylic acid, 2-naphthoic acid, 6-hydroxy, 2-hydroxy-6-naphthoic acid | |
| C1=CC2=C(C=CC(=C2)O)C=C1C(=O)O | |
| 188.182 | |
| 188.18 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (4)-0398
RTECSNumber : QN3722500
TSCA : Yes