missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy-2-naphthoic Acid 98.0+%, TCI America™

Catalog No. H06125G
Change view
Click to view available options
Quantity:
5 g
25 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06125G 5 g
H061225G 25 g
H0612500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H06125G Supplier TCI America Supplier No. H06125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16712-64-4
Molecular Formula C11H8O3
Molecular Weight (g/mol) 188.182
MDL Number MFCD00060070
InChI Key KAUQJMHLAFIZDU-UHFFFAOYSA-N
Synonym 6-hydroxy-2-naphthoic acid, 2-naphthalenecarboxylic acid, 6-hydroxy, 6-hydroxy-2-naphthalenecarboxylic acid, 6-carboxy-2-naphthol, 2-hydroxynaphthalene-6-carboxylic acid, 6-hydroxy-beta-naphthoic acid, unii-72xa858i6p, 6-hydroxy-naphthalene-2-carboxylic acid, 2-naphthoic acid, 6-hydroxy, 2-hydroxy-6-naphthoic acid
PubChem CID 85557
IUPAC Name 6-hydroxynaphthalene-2-carboxylic acid
SMILES C1=CC2=C(C=CC(=C2)O)C=C1C(=O)O

Specifications

Melting Point 248°C
Quantity 5 g
Formula Weight 188.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Hydroxy-2-naphthoic Acid
EINECSNumber (4)-0398
RTECSNumber QN3722500
TSCA Yes