Learn More
6-Hydroxy-2-naphthoic acid, 99%
CAS: 16712-64-4 | C11H8O3 | 188.182 g/mol
Supplier: Thermo Scientific Chemicals B2179106
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 6-Hydroxy-2-naphthoic acid | |
| 248°C to 250°C | |
| MFCD00060070 | |
| 2692084 | |
| KAUQJMHLAFIZDU-UHFFFAOYSA-N | |
| 6-hydroxynaphthalene-2-carboxylic acid | |
| 85557 | |
| 99% |
| 16712-64-4 | |
| C11H8O3 | |
| 5 g | |
| 6-hydroxy-2-naphthoic acid, 2-naphthalenecarboxylic acid, 6-hydroxy, 6-hydroxy-2-naphthalenecarboxylic acid, 6-carboxy-2-naphthol, 2-hydroxynaphthalene-6-carboxylic acid, 6-hydroxy-beta-naphthoic acid, unii-72xa858i6p, 6-hydroxy-naphthalene-2-carboxylic acid, 2-naphthoic acid, 6-hydroxy, 2-hydroxy-6-naphthoic acid | |
| C1=CC2=C(C=CC(=C2)O)C=C1C(=O)O | |
| 188.182 | |
| 188.18 |
Chemical Identifiers
| 16712-64-4 | |
| 188.182 | |
| KAUQJMHLAFIZDU-UHFFFAOYSA-N | |
| 85557 | |
| C1=CC2=C(C=CC(=C2)O)C=C1C(=O)O |
| C11H8O3 | |
| MFCD00060070 | |
| 6-hydroxy-2-naphthoic acid, 2-naphthalenecarboxylic acid, 6-hydroxy, 6-hydroxy-2-naphthalenecarboxylic acid, 6-carboxy-2-naphthol, 2-hydroxynaphthalene-6-carboxylic acid, 6-hydroxy-beta-naphthoic acid, unii-72xa858i6p, 6-hydroxy-naphthalene-2-carboxylic acid, 2-naphthoic acid, 6-hydroxy, 2-hydroxy-6-naphthoic acid | |
| 6-hydroxynaphthalene-2-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 240-759-7
RTECSNumber : QN3722500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only