missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy-2,4-dimethylpyrimidine, 99%

Catalog No. AAA1096006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1096006 5 g
AAA1096014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1096006 Supplier Thermo Scientific Chemicals Supplier No. A1096006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6622-92-0 | C6H8N2O | 124.143 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6622-92-0
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.143
MDL Number MFCD00006105
InChI Key UQFHLJKWYIJISA-UHFFFAOYSA-N
Synonym 2,4-dimethyl-6-hydroxypyrimidine, 2,6-dimethylpyrimidin-4-ol, 4-hydroxy-2,6-dimethylpyrimidine, 6-hydroxy-2,4-dimethylpyrimidine, 2,6-dimethyl-4-pyrimidinol, 4 1h-pyrimidinone, 2,6-dimethyl, 2,6-dimethyl-4-hydroxypyrimidine, 2,6-dimethylpyrimidin-4 3h-one, 2,4-dimethyl-6-oxypyrimidine, t6vm dnj c1 e1
PubChem CID 81098
IUPAC Name 2,6-dimethyl-1H-pyrimidin-4-one
SMILES CC1=CC(=O)N=C(N1)C

Specifications

Melting Point 198°C to 200°C
Quantity 5 g
Beilstein 114324
Formula Weight 124.14
Percent Purity 99%
Chemical Name or Material 6-Hydroxy-2,4-dimethylpyrimidine
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 229-577-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only