accessibility menu, dialog, popup

UserName

CH3ONH3CHN

CAS RN 6622-92-0

IUPAC Name: 2,6-dimethyl-1,4-dihydropyrimidin-4-one
Synonyms: 2,6-dimethyl-1H-pyrimidin-4-one
Molecular Weight (g/mol): 124.14
Molecular Formula: C6H8N2O
InChi Key: UQFHLJKWYIJISA-UHFFFAOYSA-N
SMILES: CC1=CC(=O)N=C(C)N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)