missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy-2,3-dihydrobenzo[b]furan-3-one, 97%

Catalog No. AAL1095106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1095106 5 g
AAL1095103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1095106 Supplier Thermo Scientific Chemicals Supplier No. L1095106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6272-26-0 | C8H6O3 | 150.13 g/mol

6-Hydroxy-2,3-dihydrobenzo[b]furan-3-one, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Hydroxy-2,3-dihydrobenzo[b]furan-3-one, is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 6272-26-0
Molecular Formula C8H6O3
Molecular Weight (g/mol) 150.13
MDL Number MFCD00068174
InChI Key GBDMODVZBPFQKI-UHFFFAOYSA-N
Synonym 6-hydroxybenzofuran-3 2h-one, 6-hydroxy-2h-benzofuran-3-one, 6-hydroxy-2,3-dihydrobenzo b furan-3-one, 6-hydroxy-1-benzofuran-3 2h-one, 6-hydroxybenzofuran-3-one, 3 2h-benzofuranone, 6-hydroxy, 6-hydroxy-3-coumaranone, 6-hydroxy-2,3-dihydro-1-benzofuran-3-one, 6-hydroxy-3 2h-benzofuranone, 6-hydroxy-2h-1-benzofuran-3-one
PubChem CID 235717
IUPAC Name 6-hydroxy-2,3-dihydro-1-benzofuran-3-one
SMILES OC1=CC=C2C(=O)COC2=C1

Specifications

Melting Point 240°C to 242°C
Quantity 5 g
Beilstein 121444
Solubility Information Slightly soluble in water.
Formula Weight 150.14
Percent Purity 97%
Chemical Name or Material 6-Hydroxy-2,3-dihydrobenzo[b]furan-3-one
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only