accessibility menu, dialog, popup

UserName

OHOO

CAS RN 6272-26-0

IUPAC Name: 6-hydroxy-2,3-dihydro-1-benzofuran-3-one
Synonyms: 6-hydroxy-2H-1-benzofuran-3-one
Molecular Weight (g/mol): 150.13
Molecular Formula: C8H6O3
InChi Key: GBDMODVZBPFQKI-UHFFFAOYSA-N
SMILES: OC1=CC=C2C(=O)COC2=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)