missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Hydroxy-1-tetralone 98.0+%, TCI America™
$146.31 - $579.48
Chemical Identifiers
| CAS | 3470-50-6 |
|---|---|
| Molecular Formula | C10H10O2 |
| Molecular Weight (g/mol) | 162.188 |
| MDL Number | MFCD00156670 |
| InChI Key | FNSQPQKPPGALFA-UHFFFAOYSA-N |
| Synonym | 6-hydroxy-1-tetralone, 6-hydroxy-3,4-dihydronaphthalen-1 2h-one, 6-hydroxy-3,4-dihydro-1 2h-naphthalenone, 6-hydroxy-1,2,3,4-tetrahydronaphthalen-1-one, 6-hydroxytetralone, 1-tetralon-6-ol, 6-hydroxytetralin-1-one, 2,5-dimethoxy naphthalene, 1 2h-naphthalenone, 3,4-dihydro-6-hydroxy, 6-hydroxy-2,3,4-trihydronaphthalen-1-one |
| PubChem CID | 4679494 |
| IUPAC Name | 6-hydroxy-3,4-dihydro-2H-naphthalen-1-one |
| SMILES | C1CC2=C(C=CC(=C2)O)C(=O)C1 |
Chemical Identifiers
| 3470-50-6 | |
| 162.188 | |
| FNSQPQKPPGALFA-UHFFFAOYSA-N | |
| 4679494 | |
| C1CC2=C(C=CC(=C2)O)C(=O)C1 |
| C10H10O2 | |
| MFCD00156670 | |
| 6-hydroxy-1-tetralone, 6-hydroxy-3,4-dihydronaphthalen-1 2h-one, 6-hydroxy-3,4-dihydro-1 2h-naphthalenone, 6-hydroxy-1,2,3,4-tetrahydronaphthalen-1-one, 6-hydroxytetralone, 1-tetralon-6-ol, 6-hydroxytetralin-1-one, 2,5-dimethoxy naphthalene, 1 2h-naphthalenone, 3,4-dihydro-6-hydroxy, 6-hydroxy-2,3,4-trihydronaphthalen-1-one | |
| 6-hydroxy-3,4-dihydro-2H-naphthalen-1-one |
Specifications
| 3470-50-6 | |
| C10H10O2 | |
| 5 g | |
| FNSQPQKPPGALFA-UHFFFAOYSA-N | |
| 6-hydroxy-3,4-dihydro-2H-naphthalen-1-one | |
| 4679494 | |
| ≥98.0% (GC,T) | |
| 6-Hydroxy-1-tetralone |
| 155°C | |
| MFCD00156670 | |
| 6-hydroxy-1-tetralone, 6-hydroxy-3,4-dihydronaphthalen-1 2h-one, 6-hydroxy-3,4-dihydro-1 2h-naphthalenone, 6-hydroxy-1,2,3,4-tetrahydronaphthalen-1-one, 6-hydroxytetralone, 1-tetralon-6-ol, 6-hydroxytetralin-1-one, 2,5-dimethoxy naphthalene, 1 2h-naphthalenone, 3,4-dihydro-6-hydroxy, 6-hydroxy-2,3,4-trihydronaphthalen-1-one | |
| C1CC2=C(C=CC(=C2)O)C(=O)C1 | |
| 162.188 | |
| 162.19 | |
| Crystalline Powder |
Safety and Handling
TSCA : No