missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy-1,2,3,4-tetrahydroisoquinoline 98.0+%, TCI America™

Catalog No. H15725G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15725G 5 g
H15721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15725G Supplier TCI America Supplier No. H15725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14446-24-3
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.193
MDL Number MFCD01717047
InChI Key SCMZIFSYPJICCV-UHFFFAOYSA-N
Synonym 6-hydroxy-1,2,3,4-tetrahydroisoquinoline, 1,2,3,4-tetrahydro-isoquinolin-6-ol, 1,2,3,4-tetrahydro-6-isoquinolinol, 6-isoquinolinol, 1,2,3,4-tetrahydro, pubchem13941, 6-hydroxytetrahydroisochinolin, ksc376k4j, 6-hydroxy-1,2,3,4-tetrahydro-isoquinoline
PubChem CID 26698
IUPAC Name 1,2,3,4-tetrahydroisoquinolin-6-ol
SMILES C1CNCC2=C1C=C(C=C2)O

Specifications

Melting Point 192°C
Color White-Yellow
Quantity 5 g
Formula Weight 149.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Hydroxy-1,2,3,4-tetrahydroisoquinoline
RTECSNumber NX5898000
TSCA No