accessibility menu, dialog, popup

UserName

HONH

CAS RN 14446-24-3

IUPAC Name: 1,2,3,4-tetrahydroisoquinolin-6-ol
Synonyms: 1,2,3,4-tetrahydroisoquinolin-6-ol
Molecular Weight (g/mol): 149.19
Molecular Formula: C9H11NO
InChi Key: SCMZIFSYPJICCV-UHFFFAOYSA-N
SMILES: OC1=CC=C2CNCCC2=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)