missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy-1,2,3,4-tetrahydroisoquinoline, 97%

Catalog No. p-7055833
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64512MD 250 mg
AAH6451203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH64512MD Supplier Thermo Scientific Chemicals Supplier No. H64512MD
Only null left

CAS: 14446-24-3 | C9H11NO | 149.193 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14446-24-3
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.193
MDL Number MFCD01717047
InChI Key SCMZIFSYPJICCV-UHFFFAOYSA-N
Synonym 6-hydroxy-1,2,3,4-tetrahydroisoquinoline, 1,2,3,4-tetrahydro-isoquinolin-6-ol, 1,2,3,4-tetrahydro-6-isoquinolinol, 6-isoquinolinol, 1,2,3,4-tetrahydro, pubchem13941, 6-hydroxytetrahydroisochinolin, ksc376k4j, 6-hydroxy-1,2,3,4-tetrahydro-isoquinoline
PubChem CID 26698
IUPAC Name 1,2,3,4-tetrahydroisoquinolin-6-ol
SMILES C1CNCC2=C1C=C(C=C2)O

Specifications

Melting Point 192°C to 193°C
Quantity 250 mg
Beilstein 127578
Formula Weight 149.19
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 6-Hydroxy-1,2,3,4-tetrahydroisoquinoline
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber NX5898000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only