Learn More
6-Heptynoic acid, 95%
CAS: 30964-00-2 | C7H10O2 | 126.16 g/mol
$84.27 - $1,279.46
Chemical Identifiers
| CAS | 30964-00-2 |
|---|---|
| Molecular Formula | C7H10O2 |
| Molecular Weight (g/mol) | 126.16 |
| MDL Number | MFCD00239430 |
| InChI Key | OFCPMJGTZUVUSM-UHFFFAOYSA-N |
| Synonym | 6-heptynoic acid, 6-heptynoicacid, 6-heptynoic alcohol, ksc222a4l, ofcpmjgtzuvusm-uhfffaoysa |
| PubChem CID | 4377950 |
| IUPAC Name | hept-6-ynoic acid |
| SMILES | OC(=O)CCCCC#C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC457390010
|
N/A | 1 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC457390050
|
N/A | 5 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC457390250
|
N/A | 25 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 30964-00-2 | |
| 126.16 | |
| OFCPMJGTZUVUSM-UHFFFAOYSA-N | |
| 4377950 | |
| OC(=O)CCCCC#C |
| C7H10O2 | |
| MFCD00239430 | |
| 6-heptynoic acid, 6-heptynoicacid, 6-heptynoic alcohol, ksc222a4l, ofcpmjgtzuvusm-uhfffaoysa | |
| hept-6-ynoic acid |
Specifications
| 30964-00-2 | |
| 94% min. (GC) | |
| C7H10O2 | |
| 6-heptynoic acid, 6-heptynoicacid, 6-heptynoic alcohol, ksc222a4l, ofcpmjgtzuvusm-uhfffaoysa | |
| OC(=O)CCCCC#C | |
| 126.16 | |
| 126.16 | |
| 6-Heptynoic acid |
| Authentic | |
| Glass Bottle | |
| MFCD00239430 | |
| OFCPMJGTZUVUSM-UHFFFAOYSA-N | |
| hept-6-ynoic acid | |
| 4377950 | |
| 95% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
RUO – Research Use Only