missing translation for 'onlineSavingsMsg'
Learn More

6-Heptynoic acid, 95%

Catalog No. AC457390010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC457390010 1 g
AC457390050 5 g
AC457390250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC457390010 Supplier Thermo Scientific Chemicals Supplier No. 457390010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 30964-00-2 | C7H10O2 | 126.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 30964-00-2
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.16
MDL Number MFCD00239430
InChI Key OFCPMJGTZUVUSM-UHFFFAOYSA-N
Synonym 6-heptynoic acid, 6-heptynoicacid, 6-heptynoic alcohol, ksc222a4l, ofcpmjgtzuvusm-uhfffaoysa
PubChem CID 4377950
IUPAC Name hept-6-ynoic acid
SMILES OC(=O)CCCCC#C

Specifications

Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 126.16
Percent Purity 95%
Chemical Name or Material 6-Heptynoic acid
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

RUO – Research Use Only