missing translation for 'onlineSavingsMsg'
Learn More

6-Heptynoic acid, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 457390250

 View more versions of this product

Catalog No. AC457390250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

6-Heptynoic acid
Authentic
C7H10O2
25g
OFCPMJGTZUVUSM-UHFFFAOYSA-N
hept-6-ynoic acid
4377950
95%
30964-00-2
94% min. (GC)
MFCD00239430
6-heptynoic acid, 6-heptynoicacid, 6-heptynoic alcohol, ksc222a4l, ofcpmjgtzuvusm-uhfffaoysa
OC(=O)CCCCC#C
126.16
126.16

Chemical Identifiers

30964-00-2
126.16
OFCPMJGTZUVUSM-UHFFFAOYSA-N
4377950
OC(=O)CCCCC#C
C7H10O2
MFCD00239430
6-heptynoic acid, 6-heptynoicacid, 6-heptynoic alcohol, ksc222a4l, ofcpmjgtzuvusm-uhfffaoysa
hept-6-ynoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only