missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoroveratraldehyde, 97%, Thermo Scientific™

Chemical Identifiers

CAS 71924-62-4
Molecular Formula C9H9FO3
Molecular Weight (g/mol) 184.17
MDL Number MFCD00061108
InChI Key IBBYQNVXKFMSSI-UHFFFAOYSA-N
Synonym 6-fluoroveratraldehyde, 4,5-dimethoxy-2-fluorobenzaldehyde, benzaldehyde,2-fluoro-4,5-dimethoxy, benzaldehyde, 2-fluoro-4,5-dimethoxy, pubchem2651, acmc-1bkgo, 4.5-dimethoxy-2-fluorobenzaldehyde, 2-fluoro-4,5-dimethoxy-benzaldehyde, 4,5-dimethoxy-2-fluoro benzaldehyde
PubChem CID 603595
IUPAC Name 2-fluoro-4,5-dimethoxybenzaldehyde
SMILES COC1=CC(F)=C(C=O)C=C1OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC398090010
SDS View Documents
Thermo Scientific Chemicals
398090010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC398090050
SDS View Documents
Thermo Scientific Chemicals
398090050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

71924-62-4
184.17
IBBYQNVXKFMSSI-UHFFFAOYSA-N
603595
COC1=CC(F)=C(C=O)C=C1OC
C9H9FO3
MFCD00061108
6-fluoroveratraldehyde, 4,5-dimethoxy-2-fluorobenzaldehyde, benzaldehyde,2-fluoro-4,5-dimethoxy, benzaldehyde, 2-fluoro-4,5-dimethoxy, pubchem2651, acmc-1bkgo, 4.5-dimethoxy-2-fluorobenzaldehyde, 2-fluoro-4,5-dimethoxy-benzaldehyde, 4,5-dimethoxy-2-fluoro benzaldehyde
2-fluoro-4,5-dimethoxybenzaldehyde

Specifications

71924-62-4
100.0
Glass bottle
(CH3O)2C6H2(F)CHO
1g
IBBYQNVXKFMSSI-UHFFFAOYSA-N
2-fluoro-4,5-dimethoxybenzaldehyde
603595
97%
96.0
96% min. (GC)
C9H9FO3
MFCD00061108
6-fluoroveratraldehyde, 4,5-dimethoxy-2-fluorobenzaldehyde, benzaldehyde,2-fluoro-4,5-dimethoxy, benzaldehyde, 2-fluoro-4,5-dimethoxy, pubchem2651, acmc-1bkgo, 4.5-dimethoxy-2-fluorobenzaldehyde, 2-fluoro-4,5-dimethoxy-benzaldehyde, 4,5-dimethoxy-2-fluoro benzaldehyde
COC1=CC(F)=C(C=O)C=C1OC
184.17
184.17
6-Fluoroveratraldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only