accessibility menu, dialog, popup

UserName

CAS RN 71924-62-4

IUPAC Name: 2-fluoro-4,5-dimethoxybenzaldehyde
Synonyms: 2-fluoro-4,5-dimethoxybenzaldehyde
Molecular Weight (g/mol): 184.17
Molecular Formula: C9H9FO3
InChi Key: IBBYQNVXKFMSSI-UHFFFAOYSA-N
SMILES: COC1=CC(F)=C(C=O)C=C1OC
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)