missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoroveratraldehyde, 97%

Catalog No. AC398090050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC398090050 5 g Glass bottle
AC398090010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC398090050 Supplier Thermo Scientific Chemicals Supplier No. 398090050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 71924-62-4 | C9H9FO3 | 184.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 71924-62-4
Molecular Formula C9H9FO3
Molecular Weight (g/mol) 184.17
MDL Number MFCD00061108
InChI Key IBBYQNVXKFMSSI-UHFFFAOYSA-N
Synonym 6-fluoroveratraldehyde, 4,5-dimethoxy-2-fluorobenzaldehyde, benzaldehyde,2-fluoro-4,5-dimethoxy, benzaldehyde, 2-fluoro-4,5-dimethoxy, pubchem2651, acmc-1bkgo, 4.5-dimethoxy-2-fluorobenzaldehyde, 2-fluoro-4,5-dimethoxy-benzaldehyde, 4,5-dimethoxy-2-fluoro benzaldehyde
PubChem CID 603595
IUPAC Name 2-fluoro-4,5-dimethoxybenzaldehyde
SMILES COC1=CC(F)=C(C=O)C=C1OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 98°C to 100°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H2(F)CHO
Quantity 5 g
Formula Weight 184.17
Percent Purity 97%
Chemical Name or Material 6-Fluoroveratraldehyde
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only