missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Fluoroveratraldehyde, 97+%
Description
6-Fluoroveratraldehyde is used in the preparation of 6-[18F]fluoroveratraldehyde via nucleophilic aromatic substitution by [18F]fluoride. It is also used in the preparation of key metabolites of 6-fluoro-DOPA.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Fluoroveratraldehyde is used in the preparation of 6-[18F]fluoroveratraldehyde via nucleophilic aromatic substitution by [18F]fluoride. It is also used in the preparation of key metabolites of 6-fluoro-DOPA.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases. It is sensitive to air.
6-Fluoroveratraldehyde is used in the preparation of 6-[18F]fluoroveratraldehyde via nucleophilic aromatic substitution by [18F]fluoride. It is also used in the preparation of key metabolites of 6-fluoro-DOPA.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases. It is sensitive to air.
Specifications
Specifications
| Melting Point | 98°C to 100°C |
| Quantity | 1 g |
| Beilstein | 2109343 |
| Sensitivity | Air Sensitive |
| Formula Weight | 184.17 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 6-Fluoroveratraldehyde |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 676-950-0 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only