missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoroveratraldehyde, 97+%

Catalog No. AAB2120203
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2120203 1 g
AAB2120206 5 g
AAB2120214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2120203 Supplier Thermo Scientific Chemicals Supplier No. B2120203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 71924-62-4 | C9H9FO3 | 184.17 g/mol

6-Fluoroveratraldehyde is used in the preparation of 6-[18F]fluoroveratraldehyde via nucleophilic aromatic substitution by [18F]fluoride. It is also used in the preparation of key metabolites of 6-fluoro-DOPA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Fluoroveratraldehyde is used in the preparation of 6-[18F]fluoroveratraldehyde via nucleophilic aromatic substitution by [18F]fluoride. It is also used in the preparation of key metabolites of 6-fluoro-DOPA.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases. It is sensitive to air.

Chemical Identifiers

CAS 71924-62-4
Molecular Formula C9H9FO3
Molecular Weight (g/mol) 184.17
MDL Number MFCD00061108
InChI Key IBBYQNVXKFMSSI-UHFFFAOYSA-N
Synonym 6-fluoroveratraldehyde, 4,5-dimethoxy-2-fluorobenzaldehyde, benzaldehyde,2-fluoro-4,5-dimethoxy, benzaldehyde, 2-fluoro-4,5-dimethoxy, pubchem2651, acmc-1bkgo, 4.5-dimethoxy-2-fluorobenzaldehyde, 2-fluoro-4,5-dimethoxy-benzaldehyde, 4,5-dimethoxy-2-fluoro benzaldehyde
PubChem CID 603595
IUPAC Name 2-fluoro-4,5-dimethoxybenzaldehyde
SMILES COC1=CC(F)=C(C=O)C=C1OC

Specifications

Melting Point 98°C to 100°C
Quantity 1 g
Beilstein 2109343
Sensitivity Air Sensitive
Formula Weight 184.17
Percent Purity ≥98%
Chemical Name or Material 6-Fluoroveratraldehyde
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 676-950-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only