missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoroquinoline-2-carboxaldehyde, 97%

Catalog No. p-7056878
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66052MD 250 mg
AAH6605203 1 g
AAH6605206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH66052MD Supplier Thermo Scientific Chemicals Supplier No. H66052MD

CAS: 260430-93-1 | C10H6FNO | 175.162 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 

Chemical Identifiers

CAS 260430-93-1
Molecular Formula C10H6FNO
MDL Number MFCD06751849
InChI Key JDQMJKKGPXPPOJ-UHFFFAOYSA-N
Synonym 2-quinolinecarboxaldehyde, 6-fluoro, 6-fluoro-2-quinolinecarbaldehyde, 6-fluoro-quinoline-2-carbaldehyde, 6-fluoroquinoline-2-carboxaldehyde, 4-aminomethyl-1-isoindolinone
PubChem CID 3159605
IUPAC Name 6-fluoroquinoline-2-carbaldehyde

Specifications

Melting Point 113°C to 120°C
Quantity 250 mg
Beilstein 8479194
Sensitivity Moisture sensitive
SMILES C1=CC2=C(C=CC(=N2)C=O)C=C1F
Molecular Weight (g/mol) 175.162
Formula Weight 175.16
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 6-Fluoroquinoline-2-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only