CAS RN 260430-93-1
IUPAC Name:
6-fluoroquinoline-2-carbaldehyde
Synonyms:
6-fluoroquinoline-2-carbaldehyde
Molecular Weight (g/mol):
175.16
Molecular Formula:
C10H6FNO
InChi Key:
JDQMJKKGPXPPOJ-UHFFFAOYSA-N
SMILES:
FC1=CC=C2N=C(C=O)C=CC2=C1
1
–
1
of
1
results
1
Promotions Available
6-Fluoroquinoline-2-carboxaldehyde, 97%
6-Fluoroquinoline-2-carboxaldehyde, 97%
CAS: 260430-93-1 Molecular Formula: C10H6FNO MDL Number: MFCD06751849 InChI Key: JDQMJKKGPXPPOJ-UHFFFAOYSA-N Synonym: 2-quinolinecarboxaldehyde, 6-fluoro,6-fluoro-2-quinolinecarbaldehyde,6-fluoro-quinoline-2-carbaldehyde,6-fluoroquinoline-2-carboxaldehyde,4-aminomethyl-1-isoindolinone PubChem CID: 3159605 IUPAC Name: 6-fluoroquinoline-2-carbaldehyde