missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Fluoroindole-2-carboxylic Acid 98.0+%, TCI America™
Supplier: TCI America F08351G
Specifications
| 6-Fluoroindole-2-carboxylic Acid | |
| 250°C | |
| MFCD01863162 | |
| LRTIKMXIKAOCDM-UHFFFAOYSA-N | |
| 6-fluoro-1H-indole-2-carboxylic acid | |
| 4307444 | |
| ≥98.0% (HPLC,T) |
| 3093-97-8 | |
| C9H6FNO2 | |
| 1 g | |
| OC(=O)C1=CC2=CC=C(F)C=C2N1 | |
| 179.15 | |
| 179.15 | |
| Crystalline Powder |
Chemical Identifiers
| 3093-97-8 | |
| 179.15 | |
| LRTIKMXIKAOCDM-UHFFFAOYSA-N | |
| 6-fluoro-1H-indole-2-carboxylic acid |
| C9H6FNO2 | |
| MFCD01863162 | |
| 4307444 | |
| OC(=O)C1=CC2=CC=C(F)C=C2N1 |
Safety and Handling
TSCA : No