missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoroindole-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. F0835200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0835200MG 200 mg
F08351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0835200MG Supplier TCI America Supplier No. F0835200MG

Chemical Identifiers

CAS 3093-97-8
Molecular Formula C9H6FNO2
Molecular Weight (g/mol) 179.15
MDL Number MFCD01863162
InChI Key LRTIKMXIKAOCDM-UHFFFAOYSA-N
PubChem CID 4307444
IUPAC Name 6-fluoro-1H-indole-2-carboxylic acid
SMILES OC(=O)C1=CC2=CC=C(F)C=C2N1

Specifications

Melting Point 250°C
Quantity 200 mg
Formula Weight 179.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Fluoroindole-2-carboxylic Acid