missing translation for 'onlineSavingsMsg'
Learn More

6-Fluorochromone, 97%

Catalog No. AAH3190703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3190703 1 g
AAH3190706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3190703 Supplier Thermo Scientific Chemicals Supplier No. H3190703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 105300-38-7 | C9H5FO2 | 164.135 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 105300-38-7
Molecular Formula C9H5FO2
Molecular Weight (g/mol) 164.135
MDL Number MFCD03094002
InChI Key WHIGSYZUTMYUAX-UHFFFAOYSA-N
Synonym 6-fluorochromone, 6-fluoro-4h-chromen-4-one, 6-fluoro-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one,6-fluoro, acmc-20ebjx, 6-fluoranylchromen-4-one, 6-fluoro-1-benzopyran-4-one, 6-fluoro-4-oxo-4h-1-benzopyran
PubChem CID 688892
IUPAC Name 6-fluorochromen-4-one
SMILES C1=CC2=C(C=C1F)C(=O)C=CO2

Specifications

Melting Point 166°C to 169°C
Quantity 1 g
Formula Weight 164.14
Percent Purity 97%
Chemical Name or Material 6-Fluorochromone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only