accessibility menu, dialog, popup

UserName

FOO

CAS RN 105300-38-7

IUPAC Name: 6-fluoro-4H-chromen-4-one
Synonyms: 6-fluorochromen-4-one
Molecular Weight (g/mol): 164.14
Molecular Formula: C9H5FO2
InChi Key: WHIGSYZUTMYUAX-UHFFFAOYSA-N
SMILES: FC1=CC=C2OC=CC(=O)C2=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)